Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCK-AR Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2680S100UG
This item is not returnable.
View return policy
Description
CCK-AR Monoclonal specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry.Specifications
CCK-AR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P32238 | |
CCKAR | |
NS0 mouse myeloma cell line transfected with human CCK-A R Met1-Gln428 Accession # P32238 | |
RUO | |
886 | |
Human | |
IgG2a |
Flow Cytometry | |
377251 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CCK1R, CCK1-R, CCK-A, CCK-A R, CCK-A receptor, CCKAR, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor | |
Mouse | |
100 μg | |
Primary | |
Detects human CCK-A R. Stains human CCK-A R transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction