Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCK-AR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | CCK-AR |
---|---|
Clone | 377251 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells |
Applications | Flow Cytometry |
Description
CCK-AR Monoclonal specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry.Specifications
CCK-AR | |
LYOPH | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
P32238 | |
886 | |
NS0 mouse myeloma cell line transfected with human CCK-A R Met1-Gln428 Accession # P32238 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCK-A R. Stains human CCK-A R transfectants but not irrelevant transfectants. |
377251 | |
Flow Cytometry 2.5 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCK1R, CCK1-R, CCK-A, CCK-A R, CCK-A receptor, CCKAR, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor | |
CCKAR | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title