Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL1/I-309/TCA-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF272
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF272 Supplier R&D Systems Supplier No. AF272
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

CCL1/I-309/TCA-3 Polyclonal specifically detects CCL1/I-309/TCA-3 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL1/I-309/TCA-3
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.4-2.0 ug/mL
Gene Accession No. P22362
Gene Alias C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3
Gene Symbols CCL1
Host Species Goat
Immunogen E. coli-derived recombinant human CCL1/I-309/TCA-3 Lys24-Lys96 Accession # P22362
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6346
Test Specificity Detects human CCL1/I-309/TCA-3 in direct ELISAs and Western blots. Neutralizes the biological activity of recombinant human CCL1. Does not block the biological activity of recombinant mouse TCA-3.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.