Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL11/Eotaxin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF320SP
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Polyclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CCL11/Eotaxin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
Goat | |
Affinity Purified | |
RUO | |
6356 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.5-2.5 ug/mL | |
Q6I9T4 | |
CCL11 | |
E. coli-derived recombinant human CCL11/Eotaxin, Gly24-Pro97, Accession # Q6I9T4 | |
25 μg | |
Primary | |
Detects human CCL11/Eotaxin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse CCL11/Eotaxin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction