Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL13/MCP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB327SP
This item is not returnable.
View return policy
Description
CCL13/MCP-4 Monoclonal specifically detects CCL13/MCP-4 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
CCL13/MCP-4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
6357 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Neutralization, Flow Cytometry, CyTOF, ELISA | |
73506 | |
Neutralization 10-30 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q99616.1 | |
CCL13 | |
E. coli-derived recombinant human CCL13/MCP-4 Gln24-Thr98 Accession # Q99616.1 | |
25 μg | |
Primary | |
Detects human CCL13/MCP-4 in ELISAs. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human (rh) CCL2 is observed and no significant cross-reactivity or interference with rhCCL5, 7, 8, 11, 24, recombinant mouse CCL2, 5, 7, 11 or 12 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction