Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL13/MCP-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF327 Supplier R&D Systems Supplier No. AF327
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

CCL13/MCP-4 Polyclonal specifically detects CCL13/MCP-4 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL13/MCP-4
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 15-40 ug/mL
Gene Accession No. Q99616
Gene Alias C-C motif chemokine 13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13
Gene Symbols CCL13
Host Species Goat
Immunogen E. coli-derived recombinant human CCL13/MCP-4, Gln24-Thr98, Accession # Q99616
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6357
Test Specificity Detects human CCL13/MCP-4 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.