Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL14/HCC-1/HCC-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3241R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3241R100 100 μg
MAB3241RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3241R100 Supplier R&D Systems Supplier No. MAB3241R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL14/HCC-1/HCC-3 Monoclonal specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL14/HCC-1/HCC-3
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 256409R
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_116738
Gene Alias C-C motif chemokine 14, CCL14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14
Host Species Rat
Immunogen E. coli-derived recombinant human CCL14/HCC-1/HCC-3 Thr20-Asn109 Accession # NP_116738
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6358
Test Specificity Detects human CCL14/HCC-1/HCC-3 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.