Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL15/MIP-1 delta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF363SP
This item is not returnable.
View return policy
Description
CCL15/MIP-1 delta Polyclonal specifically detects CCL15/MIP-1 delta in Human samples. It is validated for Western Blot, Neutralization.Specifications
CCL15/MIP-1 delta | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP1 delta, MIP-1 delta, MIP5, MIP-5, MIP-5MIP-1d, Mrp-2b, NCC-3, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15 | |
Goat | |
Affinity Purified | |
RUO | |
6359 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 1-5 ug/mL | |
Q16663 | |
CCL15 | |
E. coli-derived recombinant human CCL15/MIP-1 delta Gln22-Ile113 Accession # Q16663 | |
25 μg | |
Primary | |
Detects human CCL15/MIP-1 delta in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) MPIF-1 is observed and less than 1% cross-reactivity with rhMIP-1 alpha is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction