Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL17/TARC Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $690.00
Specifications
Antigen | CCL17/TARC |
---|---|
Clone | 54015 |
Dilution | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Description
CCL17/TARC Monoclonal specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
CCL17/TARC | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
CCL17 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
54015 | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q92583.1 | |
6361 | |
E. coli-derived recombinant human CCL17/TARC Ala24-Ser94 Accession # Q92583.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCL17/TARC in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9, 11, 12, 17, 19, 20, 21, 22, 24, 25, 27 or recombinant rat CCL20 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title