Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL2/JE/MCP-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC279G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC279G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC279G025 Supplier R&D Systems Supplier No. IC279G025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL2/JE/MCP-1
Applications Flow Cytometry
Classification Monoclonal
Clone 24822
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P13500
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Gene Symbols CCL2
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL2/JE/MCP-1 Accession # P13500
Purification Method Protein A or G purified from ascites
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Detects human CCL2/JE/MCP-1 in Western blots. Does not cross-react with recombinant human CCL1, 3, 4, 5, 7, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 9,10, MIP-1 gamma , 11, 12, 17, 19, 20, 22, 24, 25, or recombinant rat CCL20.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.