Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL2/JE/MCP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB679R100
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CCL2/JE/MCP-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CCL2/JE/MCP-1 in direct ELISAs. | |
Human | |
Lyophilized |
Western Blot, Neutralization, ELISA | |
23007R | |
Western Blot 1 μg;/mL, Neutralization 0.5-2.0 μg;/mL, ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
P13500 | |
CCL2 | |
E. coli-derived recombinant human CCL2/JE/MCP-1, Gln24-Thr99, Accession No. P13500 | |
100 μg | |
6347.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction