Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL2/JE/MCP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF279NA
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Polyclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CCL2/JE/MCP-1 | |
Polyclonal | |
Unconjugated | |
P13500 | |
CCL2 | |
E. coli-derived recombinant human CCL2/JE/MCP-1 Gln24-Thr99 Accession # P13500 | |
100 μg | |
Primary | |
Detects human CCL2/JE/MCP-1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 10-25 ug/mL, Neutralization 1.5-4.5 ug/mL | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
Goat | |
Antigen Affinity-purified | |
RUO | |
6347 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction