Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL2/JE/MCP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB279500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB279500 500 μg
MAB279SP 25 μg
MAB279100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB279500 Supplier R&D Systems Supplier No. MAB279500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 14 publications

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL2/JE/MCP-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 24822
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.9-4.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Gene Symbols CCL2
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL2/JE/MCP-1
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Detects human CCL2/JE/MCP-1 in Western blots. Does not cross-react with recombinant human (rh) CCL1, 3, 4, 5, 7, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, rmCCL1, 2, 3, 4, 5, 6, 7, 9/10MIP-1 gamma , 11, 12, 17, 19, 20, 22, 24, 25, or rrCCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.