Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Description
The Human CCL2/MCP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL2/JE/MCP-1 in cell culture supernates (200 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).
Specifications
Specifications
Assay Range | 31.20 - 2000pg/mL |
Assay Sensitivity | 10pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
Specificity | Natural and Recombinant Human MCP-1 |
Sufficient For | 1 x 96-well Plate |
Target Species | Human |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction