Learn More
Description
Specifications
Specifications
| Assay Sensitivity | The minimum detectable dose of human MIP3a was determined to be approximately 0.2ng/mL. |
| Product Type | ELISA Kit |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 6364 |
| Gene Symbol | CCL20 |
| Kit Contents | Capture Antibody: 0.5mg/mL of mouse anti-MIP3 alpha monoclonal antibody (clone 319F6), Detection Antibody: 0.5mg/mL of HRPO-conjugated anti-MIP3 alpha mouse monoclonal antibody (clone 206D9), Standard: Each vial contains 1 g/mL of recombinant human MIP3 alpha produced, purified and concentrated from eukaryotic cells. A 7-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20ng/mL is recommende |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
