Learn More
Human CCL21/6Ckine Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 15 publications
Supplier:  R&D Systems AF366SP
Catalog No. AF366SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CCL21/6Ckine Polyclonal specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
Specifications
CCL21/6Ckine
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4
Goat
Affinity Purified
RUO
6366
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization, Immunocytochemistry
LYOPH
Western Blot 0.1 ug/mL, Neutralization 1-4 ug/mL, Immunocytochemistry 5-15 ug/mL
Q6ICR7
CCL21
E. coli-derived recombinant human CCL21/6Ckine Ser24-Pro134 Accession # Q6ICR7
25 μg
Primary
Detects human CCL21/6Ckine in direct ELISAs and Western blots.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only