Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL21/6Ckine Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3661SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3661SP 25 μg
MAB3661 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3661SP Supplier R&D Systems Supplier No. MAB3661SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL21/6Ckine
Applications Western Blot, ELISA
Classification Monoclonal
Clone 54111
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00585
Gene Alias 6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4
Gene Symbols CCL21
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL21/6Ckine Ser24-Pro134 Accession # O00585
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6366
Test Specificity Detects human CCL21/6Ckine in ELISAs and Western blots. In Western blots, shows 100% cross-reactivity with recombinant human (rh) CCL24. Under reducing conditions, shows approximately 10-50% cross-reactivity with rhCCL2, 8, 13, 28, and rmCCL2. Does not cross-react with rhCCL1, 3, 4, 5, 7, 9, 11, 14a, 15, 17, 18, 19, 20, 22, 23, 25, rmCCL1, 3, 4, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, 25, 28, or rrCCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.