Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL22/MDC PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems IC3361P
This item is not returnable.
View return policy
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CCL22/MDC | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6367 | |
Human | |
Purified |
Flow Cytometry | |
57203 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
O00626.1 | |
CCL22 | |
E. coli-derived recombinant human CCL22/MDC Gly25-Gln93 Accession # O00626.1 | |
100 Tests | |
Primary | |
Detects human CCL22/MDC in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 9/10/MIP-1 gamma , 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction