Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL23/Ck beta 8-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF508
This item is not returnable.
View return policy
Description
CCL23/Ck beta 8-1/MIP3 Polyclonal specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot, Neutralization.Specifications
CCL23/Ck beta 8-1/MIP3 | |
Polyclonal | |
Unconjugated | |
AAD10846 | |
CCL23 | |
E. coli-derived recombinant human CCL23/Ck beta 8-1 Met46-Asn137 Accession # AAD10846 | |
100 μg | |
Primary | |
Detects CCL23/Ck beta 8-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human MPIF-1 is observed. Additionally, in direct ELISAs, no cross-reactivity with _______ is observed. Neutralizes the biological activity of human CCL23/Ck beta 8-1. It will also neutralize the biological activity of recombinant human MPIF-1 at a 10-fold higher Ig concentration. | |
Human | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.15-0.75 ug/mL | |
C6 beta-chemokine, chemokine (C-C motif) ligand 23, Ck beta 8-1, CKb8, CKB-8, Ckb81, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23 | |
Goat | |
Affinity Purified | |
RUO | |
6368 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction