Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL23/Ck beta 8-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF508
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF508 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF508 Supplier R&D Systems Supplier No. BAF508
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CCL23/Ck beta 8-1/MIP3 Polyclonal specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen CCL23/Ck beta 8-1/MIP3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. AAD10846
Gene Alias C6 beta-chemokine, chemokine (C-C motif) ligand 23, Ck beta 8-1, CKb8, CKB-8, Ckb81, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Gene Symbols CCL23
Host Species Goat
Immunogen E. coli-derived recombinant human CCL23/CK beta 8-1 Met46-Asn137 Accession # AAD10846
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6368
Test Specificity Detects human CCL23/Ck beta 8-1 in ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.