Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL23/MPIF-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF371SP
This item is not returnable.
View return policy
Description
CCL23/MPIF-1 Polyclonal specifically detects CCL23/MPIF-1 in Human samples. It is validated for Western Blot, Neutralization.Specifications
CCL23/MPIF-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCL23 | |
E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773 | |
25 μg | |
Primary | |
Detects human CCL23/MPIF-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) MIP-1 delta , rhHCC-1, rhMIP-1 alpha and recombinant mouse MIP-1 gamma is observed. | |
Human | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
P55773 | |
Goat | |
Affinity Purified | |
RUO | |
6368 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction