Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL25/TECK Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF334
This item is not returnable.
View return policy
Description
CCL25/TECK Polyclonal specifically detects CCL25/TECK in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
| CCL25/TECK | |
| Polyclonal | |
| Biotin | |
| AAB69981 | |
| CCL25 | |
| E. coli-derived recombinant human CCL25/TECK Gln24-Leu150 Accession # AAB69981 | |
| 50 μg | |
| Primary | |
| Detects human CCL25/TECK in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with rmTECK is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6370 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction