Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL26/Eotaxin-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6532SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6532SP 25 μg
MAB6532100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6532SP Supplier R&D Systems Supplier No. MAB6532SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL26/Eotaxin-3 Monoclonal specifically detects CCL26/Eotaxin-3 in Human samples. It is validated for ELISA.

Specifications

Antigen CCL26/Eotaxin-3
Applications ELISA
Classification Monoclonal
Clone 115005
Conjugate Unconjugated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9Y258.1
Gene Alias CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1
Host Species Mouse
Immunogen E. coli-derived human CCL26/Eotaxin-3 protein, Thr24-Leu94, Accession # Q9Y258.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10344
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.