Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL26/Eotaxin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | CCL26/Eotaxin-3 |
---|---|
Clone | 115009 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6533SP
![]() |
R&D Systems
MAB6533SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6533100
![]() |
R&D Systems
MAB6533100 |
100 μg |
Each for $432.50
|
|
|||||
Description
CCL26/Eotaxin-3 Monoclonal specifically detects CCL26/Eotaxin-3 in Human samples. It is validated for ELISA.Specifications
CCL26/Eotaxin-3 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9Y258.1 | |
10344 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
115009 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1 | |
E. coli-derived human CCL26/Eotaxin-3, Thr24-Leu94, Accession # Q9Y258.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title