Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL27/CTACK Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3762
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3762 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3762 Supplier R&D Systems Supplier No. MAB3762
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL27/CTACK
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 124315
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1.5-5.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_006655
Gene Alias ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27
Gene Symbols CCL27
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL27/CTACK Phe25-Gly112 Accession # NP_006655
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10850
Test Specificity Detects human CCL27/CTACK in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) CCL27CTACK is observed, 10% cross-reactivity with recombinant human (rh) CCL28 is observed, and no cross-reactivity with rhCCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, rmCCL1, 2, 3, 4, 5, 6, 7, 9/10, 11, 12, 17, 19, 20, 21, 22, 24, 25, 28, or recombinant rat CCL20 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.