Learn More
Human CCL27/CTACK Biotinylated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCL27/CTACK |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL |
| Gene Accession No. | Q9Y4X3 |
| Gene Alias | ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 |
| Gene Symbols | CCL27 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.