Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL3/CCL4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB270SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB270SP 25 μg
MAB270100 100 μg
MAB270500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB270SP Supplier R&D Systems Supplier No. MAB270SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.

Specifications

Antigen CCL3/CCL4
Applications Western Blot, Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 93321
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.075-0.375 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10147
Gene Symbols CCL3
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects human CCL3/MIP-1 alpha in direct ELISAs and Western blots. In direct ELISAs, greater than 75% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed while approximately 30% cross-reactivity with recombinant mouse (rm) CCL3, rmCCL4/MIP-1 beta , and rhCCL14 is observed. No cross-reactivity with rhCCL1, 2, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, rmCCL1, 2, 5, 6, 7, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.