Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL3/CCL4 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2701A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2701A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2701A Supplier R&D Systems Supplier No. IC2701A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL3/CCL4
Applications Flow Cytometry
Classification Monoclonal
Clone 93342
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P10147
Gene Symbols CCL3
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects human CCL3/MIP-1 alpha in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed. Does not cross-react with rhCCL1, 2, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 2, 3, 5, 6, 7, CCL9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.