Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL3/MIP-1 alpha Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2701T100UG
This item is not returnable.
View return policy
Description
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CCL3/CCL4 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P10147 | |
CCL3 | |
E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147 | |
100 μg | |
Primary | |
Detects human CCL3/MIP-1 alpha in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed. Does not cross-react with rhCCL1, 2, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 2, 3, 5, 6, 7, CCL9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
93342 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
6348 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction