Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL3/MIP-1 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $2020.00
Specifications
Antigen | CCL3/MIP-1 alpha |
---|---|
Clone | 14215 |
Dilution | Neutralization 3-10 ug/mL |
Applications | Neutralization |
Classification | Monoclonal |
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for ELISA, Neutralization.Specifications
CCL3/MIP-1 alpha | |
Neutralization 3-10 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
CCL3 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
14215 | |
Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6348 | |
E. coli-derived recombinant human CCL3/MIP-1 alpha | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCL3/MIP-1 alpha . |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title