Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL3/MIP-1 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB9849100
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for ELISA.Specifications
| CCL3/MIP-1alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CCL3/MIP-1 alpha in direct ELISAs. | |
| Human | |
| Supernatant |
| ELISA | |
| 93333R | |
| ELISA | |
| P10147 | |
| CCL3 | |
| E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147 | |
| 100 μg | |
| 6348 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction