Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL3/MIP-1 alpha Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Supplier: R&D Systems BAF270
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Polyclonal specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).Specifications
CCL3/MIP-1 alpha | |
Polyclonal | |
Biotin | |
P10147 | |
CCL3 | |
E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147 | |
50 μg | |
Primary | |
Detects human CCL3/MIP-1 alpha in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human (rh) MIP-1 beta , rhMIP-1 delta , rhMIP-3 alpha , rhMIP-3 beta , and recombinant mouse MIP-1 alpha is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
Goat | |
Affinity Purified | |
RUO | |
6348 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction