Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL4/MIP-1 beta Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems BAF271
This item is not returnable.
View return policy
Description
CCL4/MIP-1 beta Polyclonal specifically detects CCL4/MIP-1 beta in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).Specifications
CCL4/MIP-1 beta | |
Polyclonal | |
Biotin | |
ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2 | |
Goat | |
Affinity Purified | |
RUO | |
6351 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CCL4 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CCL4/MIP-1 beta | |
50 μg | |
Primary | |
Detects human CCL4/MIP-1 beta in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human (rh) MIP-1 alpha is observed and less than 0.1% cross-reactivity with recombinant viral (rv) MIP-I, rvMIP-II, rhMIP-1 delta , recombinant mouse (rm) MIP-1 alpha , rmMIP-1 beta , rmMIP-1 gamma , rhGRO beta , rhGRO gamma , rhMCP-1, and rhRANTES is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction