Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL4/MIP-1 beta DuoSet ELISA

Catalog No. DY27105
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY27105 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY27105 Supplier R&D Systems™ Supplier No. DY27105
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Human MIP-1beta Capture Antibody; Human MIP-1beta Detection Antibody; Human MIP-1beta Standard; Streptavidin-HRP

Human CCL4/MIP-1 beta DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL4/MIP-1 beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CCL4/MIP-1 beta.

Specifications

Quantity 5 Plates
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.