Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL4/MIP-1 beta MAb (Clone 24006), R&D Systems™
GREENER_CHOICE

Catalog No. MAB271500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB271500 500 μg
MAB271SP 25 μg
MAB271100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB271500 Supplier R&D Systems Supplier No. MAB271500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 17 publications

CCL4/MIP-1 beta Monoclonal specifically detects CCL4/MIP-1 beta in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL4/MIP-1 beta
Applications Western Blot, Neutralization, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 24006
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.3-1.2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13236
Gene Alias ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Gene Symbols CCL4
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CCL4/MIP-1 beta Ala24-Asn92 Accession # P13236
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6351
Test Specificity Detects human CCL4/MIP-1 beta in ELISAs and Western blots. In Western blots, less than 4% cross-reactivity with recombinant human (rh) CCL3/MIP-1 alpha and no cross-reactivity with recombinant mouse (rm) CCL3/MIP-1 alpha , rmCCL4/MIP-1 beta , rhCXCL8/IL-8, rhCCL5, rhCXCL1, rhCXCL2, or rhCXCL3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.