Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL7/MCP-3/MARC Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC282R100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC282R100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC282R100UG Supplier R&D Systems Supplier No. IC282R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL7/MCP-3/MARC
Applications Flow Cytometry
Classification Monoclonal
Clone 36320
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q7Z7Q8
Gene Alias C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP-3, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Gene Symbols CCL7
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL7/MCP-3/MARC Gln34-109 Accession # Q7Z7Q8
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6354
Test Specificity Detects human CCL7/MCP-3/MARC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) CCL2/MCP-1, rhCCL8/MCP-2, or rhCCL13/MCP-4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.