Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL7/MCP-3/MARC Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF282NA
This item is not returnable.
View return policy
Description
CCL7/MCP-3/MARC Polyclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot, Neutralization.Specifications
CCL7/MCP-3/MARC | |
Polyclonal | |
Unconjugated | |
Q7Z7Q8 | |
CCL7 | |
E. coli-derived recombinant human CCL7/MCP-3/MARC (R&D Systems, Catalog # 282-P3) Gln34-Leu109 Accession # Q7Z7Q8 | |
100 μg | |
Primary | |
Detects human CCL7/MCP-3/MARC in direct ELISAs and Western blots. In these formats, less than 20% cross-reactivity with recombinant human (rh) Eotaxin is observed and less than 5% cross-reactivity with rhMCP-2, rhMCP-1, and recombinant mouse JE is observed. | |
Human | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL | |
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP-3, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
Goat | |
Affinity Purified | |
RUO | |
6354 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction