Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL8/MCP-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF281NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF281NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF281NA Supplier R&D Systems Supplier No. AF281NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CCL8/MCP-2 Polyclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL8/MCP-2
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 1.5-5.0 ug/mL
Gene Accession No. P80075
Gene Alias C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Gene Symbols CCL8
Host Species Goat
Immunogen E. coli-derived recombinant human CCL8/MCP-2 Gln24-Pro99 Accession # P80075
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6355
Test Specificity Detects human CCL8/MCP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 40% cross-reactivity with recombinant human (rh) HCC-1 is observed, less than 10% cross-reactivity with rhEotaxin is observed, and less than 5% cross-reactivity with rhMCP-1 is observed. Neutralizes the biological activity of rhCCL8. This antibody will not neutralize the biological activity of rhMCP-1, recombinant mouse JE, or rhMCP-3.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.