Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR1 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB145RT100UG
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry.Specifications
CCR1 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
Swiss 3T3 mouse cell line transfected with human CCR1 Accession # P32246 | |
100 μg | |
1230 | |
Human | |
Supernatant |
Flow Cytometry | |
53504R | |
Flow Cytometry 0.25-1μg/106 cells | |
P32246 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CCR1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction