Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 23 publications
Supplier: R&D Systems MAB145SP
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CCR1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P32246 | |
CCR1 | |
Swiss 3T3 mouse cell line transfected with human CCR1 Accession # P32246 | |
25 μg | |
Primary | |
Detects human CCR1. Reacts specifically with human CCR1 transfectants and not the parental cell line in flow cytometry. No cross-reactivity with CCR5 transfectants is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
53504 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1230 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction