Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR10 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
$257.00 - $514.50
Specifications
Antigen | CCR10/GPR2 |
---|---|
Clone | 314305 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3478G025
![]() |
R&D Systems
FAB3478G025 |
25 Tests |
Each for $257.00
|
|
|||||
FAB3478G100
![]() |
R&D Systems
FAB3478G100 |
100 Tests |
Each for $514.50
|
|
|||||
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry.Specifications
CCR10/GPR2 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
CCR10 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CCR10. Stains human CCR10-transfected cells but not irrelevant transfectants. |
314305 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Human | |
P46092 | |
2826 | |
Y3 rat myeloid cell line transfected with human CCR10 Gln8-Asn362 Accession # P46092 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title