Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR11 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6899N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6899N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6899N100 Supplier R&D Systems Supplier No. FAB6899N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR11 Monoclonal specifically detects CCR11 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR11
Applications Flow Cytometry
Classification Monoclonal
Clone 674144
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9NPB9
Gene Alias ACKR4, CC chemokine receptor-like 1, C-C CKR-11, CCBP2CCX CKR, CC-CKR-11, CCR11CCR10, CCRL1, CCX-CKR, chemokine (C-C motif) receptor-like 1, orphan seven-transmembrane receptor, chemokine related, PPR1, PPR1cc motif, receptor-like protein 1, VSHK1, VSHK1CKR-11
Gene Symbols CCRL1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCR11 Accession # Q9NPB9
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51554
Test Specificity Detects human CCR11 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgM
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.