Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR11 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6899P100
This item is not returnable.
View return policy
Description
CCR11 Monoclonal specifically detects CCR11 in Human samples. It is validated for Flow Cytometry.Specifications
CCR11 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and sodium azide | |
ACKR4, CC chemokine receptor-like 1, C-C CKR-11, CCBP2CCX CKR, CC-CKR-11, CCR11CCR10, CCRL1, CCX-CKR, chemokine (C-C motif) receptor-like 1, orphan seven-transmembrane receptor, chemokine related, PPR1, PPR1cc motif, receptor-like protein 1, VSHK1, VSHK1CKR-11 | |
NS0 mouse myeloma cell line transfected with human CCR11, Accession No. Q9NPB9 | |
100 Tests | |
51554.0 | |
Human | |
Supernatant |
Flow Cytometry | |
674144 | |
Flow Cytometry 10 uL/10^6 cells | |
Q9NPB9 | |
Mouse | |
IgM-specific Affinity-purified from hybridoma culture supernatant | |
Primary | |
Detects human CCR11 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction