Learn More
Human CCR2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCR2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 48607 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Gene Accession No. | NP_001116868 |
| Gene Alias | C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor |
| Gene Symbols | CCR2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.