Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR2 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems FAB151B
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Flow Cytometry.Specifications
CCR2 | |
Monoclonal | |
Biotin | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
729230 | |
Human | |
Purified |
Flow Cytometry | |
48607 | |
Flow Cytometry 10 uL/10^6 cells | |
NP_001116868 | |
CCR2 | |
NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 | |
100 Tests | |
Primary | |
Detects human CCR2 transfectants but not the parental cell line or CCR5 transfectants. | |
Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction