Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR3 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB155C025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB155C025 25 Tests
FAB155C100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB155C025 Supplier R&D Systems Supplier No. FAB155C025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR3
Applications Flow Cytometry
Classification Monoclonal
Clone 61828
Conjugate PerCP
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P51677
Gene Alias b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Gene Symbols CCR3
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CCR3 Accession # P51677
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1232
Test Specificity Detects human CCR3 transfectants in flow cytometry. In flow cytometry, no cross-reactivity with CCR1, CCR2, CCR5, and CCR6 transfectants or the parental cell line is observed. Detects CCR3 on peripheral blood eosinophils.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.