Learn More
Human CCR4 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB1567RV100UG
Catalog No. FB1567RV100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR4
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Human CCR4 transfectants, Met1-Leu360, Accession No. P51679
100 μg
1233.0
Human
Supernatant
Flow Cytometry
205410R
Flow Cytometry 0.25-1 μg/106 cells
P51679
Mouse
Protein A or G purified from cell culture supernatant
Primary
Detects human CCR4 in direct ELISAs
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.