Learn More
Human CCR5 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCR5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CTC5R |
| Conjugate | Alexa Fluor 750 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 |
| Host Species | Mouse |
| Immunogen | CHO Chinese hamster ovary cell line transfected with human CCR5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.