Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR5 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$151.20 - $385.50
Specifications
Antigen | CCR5 |
---|---|
Clone | 45531 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB182F025
![]() |
R&D Systems
FAB182F025 |
25 Tests |
Each for $151.20
|
|
|||||
FAB182F100
![]() |
R&D Systems
FAB182F100 |
100 Tests |
Each for $385.50
|
|
|||||
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.Specifications
CCR5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
CCR5 | |
IgG2b | |
Protein A or G purified from ascites | |
Detects human CCR5. |
45531 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
P51681 | |
1234 | |
NS0 mouse myeloma cell line transfected with human CCR5 Met1-Leu352 Accession # P51681 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title