Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR6 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB195RU100UG
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry.Specifications
CCR6 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
NS0 mouse myeloma cell line transfected with human CCR6 Met1-Met374 Accession # P51684 | |
100 μg | |
1235 | |
Human | |
Supernatant |
Flow Cytometry | |
53103R | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
P51684 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CCR6. Stains human CCR6 transfectants but not the parental cell line in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction